Learn More
ATCC Clostridium sporogenes; Strain NCA 3679
Catalog No. 502384474
Encompass
Used for sterility testing and as a steam sterilization control.
Catalog No. 50-238-4474 Supplier ATCC Supplier No. 7955
Used for sterility testing and as a steam sterilization control.