Learn More
Chemglass Life Sciences Closure, 25mm, Stainless Steel
SDP
Catalog No. 501532234
Encompass_Preferred
Closure, 25mm, Stainless Steel . These closures are made from 305 Stainless Steel . They are ideal for use with culture tubes and flasks utilizing Delong Necks. The closure is designed with finger grips to allow it to fit firmly on any Delong Neck. It will withstand both dry heat sterilization and repeated autoclaving.
Catalog No. 50-153-2234 Supplier Chemglass Life Sciences Supplier No. CLS1502025
Closure, 25mm, Stainless Steel . These closures are made from 305 Stainless Steel . They are ideal for use with culture tubes and flasks utilizing Delong Necks. The closure is designed with finger grips to allow it to fit firmly on any Delong Neck. It will withstand both dry heat sterilization and repeated autoclaving.