Learn More
Labcon North America Flat Style Caps for 50mL Centrifuge Tubes, in Bags
Catalog No. 50101724
Encompass_Preferred
Cap, Flat style screw cap for Labcon 50mL centrifuge tubes; 50mL; Labcon; Endotoxin Free (non-pyrogenic); Polyethylene USP Classs VI resin; Not autoclavable; Teal color; 500 per bag/4 bags per case; Made in USA
Catalog No. 50-101-724 Supplier Labcon North America Supplier No. 30758700089
Cap, Flat style screw cap for Labcon 50mL centrifuge tubes; 50mL; Labcon; Endotoxin Free (non-pyrogenic); Polyethylene USP Classs VI resin; Not autoclavable; Teal color; 500 per bag/4 bags per case; Made in USA