Learn More
Chemglass Life Sciences Certified Bonded Closure, 9mm, Black, Natural PTFE/Off-White Silicone Pre-Slit Ultra Low Bleed Septa, for LC/MS and GC/MS
SDP
Catalog No. 501532574
Encompass_Preferred
Certified Bonded Closure, 9mm, Black, Natural PTFE/Off-White Silicone Pre-Slit Ultra Low Bleed Septa, for LC/MS and GC/MS. Certified Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.
Catalog No. 50-153-2574 Supplier Chemglass Life Sciences Supplier No. CV3840CERT
Certified Bonded Closure, 9mm, Black, Natural PTFE/Off-White Silicone Pre-Slit Ultra Low Bleed Septa, for LC/MS and GC/MS. Certified Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.