Learn More
Chemglass Life Sciences Closure Only, 9mm, Clear, Top Seal, 10mil Molded Septa
SDP
Catalog No. 501532568
Encompass_Preferred
Closure Only, 9mm, Clear, Top Seal, 10mil Molded Septa. Designed for use with Shimadzu, Spectra-Physics, Varian and other autosamplers. The Top Seal closure with a 0.010 molded septa allows for easy needle penetration and is inert to most solvents. The one-piece design saves time by eliminating liners in the closures.
Catalog No. 50-153-2568 Supplier Chemglass Life Sciences Supplier No. CV37600009
Closure Only, 9mm, Clear, Top Seal, 10mil Molded Septa. Designed for use with Shimadzu, Spectra-Physics, Varian and other autosamplers. The Top Seal closure with a 0.010 molded septa allows for easy needle penetration and is inert to most solvents. The one-piece design saves time by eliminating liners in the closures.