Learn More
Chemglass Life Sciences Bonded Closure, Standard, 9mm, Red, Open Hole, 0.40" PTFE/Silicone
SDP
Catalog No. 501311397
Encompass_Preferred
Bonded Closure, Standard, 9mm, Red, Open Hole, 0.40" PTFE/Silicone. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Non-slit design.
Catalog No. 50-131-1397 Supplier Chemglass Life Sciences Supplier No. CV3845R009
Bonded Closure, Standard, 9mm, Red, Open Hole, 0.40" PTFE/Silicone. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Non-slit design.