Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Blue, 10-425, 0.050" PTFE/Silicone/PTFE
SDP
Catalog No. 501532624
Encompass_Preferred
Preassembled Closure and Septa, Blue, 10-425, 0.050" PTFE/Silicone/PTFE. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-153-2624 Supplier Chemglass Life Sciences Supplier No. CV5075B010
Preassembled Closure and Septa, Blue, 10-425, 0.050" PTFE/Silicone/PTFE. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.