Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Black, GPI 13-425, 0.40" PTFE/Silicone/PTFE Septa
SDP
Catalog No. 501532623
Encompass_Preferred
Preassembled Closure and Septa, Black, GPI 13-425, 0.40" PTFE/Silicone/PTFE Septa. Pre-assembled closure and septa minimize contamination from handling.
Catalog No. 50-153-2623 Supplier Chemglass Life Sciences Supplier No. CV50750013
Preassembled Closure and Septa, Black, GPI 13-425, 0.40" PTFE/Silicone/PTFE Septa. Pre-assembled closure and septa minimize contamination from handling.