Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Black, GPI 13-425, .065" Red PTFE/Silicone Septa
SDP
Catalog No. 501532643
Encompass_Preferred
Preassembled Closure and Septa, Black, GPI 13-425, .065" Red PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling.
Catalog No. 50-153-2643 Supplier Chemglass Life Sciences Supplier No. CV52100013
Preassembled Closure and Septa, Black, GPI 13-425, .065" Red PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling.