Learn More
Med Vet International 3M Steri-Strip Skin Closure Strip, 1/2 X 4 inch, Nonwoven Material, Reinforced Strip, White, R1547, 50/Box
Catalog No. 502830066
Encompass_Preferred
These 1/2-inch x 4-inch Steri-Strip™ adhesive skin closures contain polymer filaments for a secure closure They have a simple design that reduces scarring and chance of infection Made using a hypoallergenic adhesive, these 3M™ Steri-Strip™ reinforced skin closure strips are safe for sensitive skin A strong hold reduces tissue damage and allows for improved cosmetic results The breathable fabric stays gentle against the skin These non-woven reinforced strips are suitable for cuts, surgical incisions and after procedures requiring sutures and staples
Catalog No. 50-283-0066 Supplier Med Vet International Supplier No. 5785BX
These 1/2-inch x 4-inch Steri-Strip™ adhesive skin closures contain polymer filaments for a secure closure They have a simple design that reduces scarring and chance of infection Made using a hypoallergenic adhesive, these 3M™ Steri-Strip™ reinforced skin closure strips are safe for sensitive skin A strong hold reduces tissue damage and allows for improved cosmetic results The breathable fabric stays gentle against the skin These non-woven reinforced strips are suitable for cuts, surgical incisions and after procedures requiring sutures and staples