Learn More
Med Vet International 3M Steri-Strip Skin Closure Strip, 1/4 X 3 inch, Nonwoven Material, Flexible Strip, Tan, B1551, 200/Case
Catalog No. 502831484
Encompass_Preferred
3M™ Steri-Strip™ blend tone skin closures have a beige color that blends with the skin, making them less visible The non-woven backing is soft on the skin for added comfort The pressure-sensitive adhesive allows for an easy application The 1/4-inch x 3-inch strips minimize tissue trauma to reduce scarring They are latex-free and are gentle on sensitive skin
Catalog No. 50-283-1484 Supplier Med Vet International Supplier No. 5786CS
3M™ Steri-Strip™ blend tone skin closures have a beige color that blends with the skin, making them less visible The non-woven backing is soft on the skin for added comfort The pressure-sensitive adhesive allows for an easy application The 1/4-inch x 3-inch strips minimize tissue trauma to reduce scarring They are latex-free and are gentle on sensitive skin