Learn More
Med Vet International Skin Closure Strip Steri-Strip 1/4 X 3 inch Nonwoven Material Reinforced Strip White, R1541, 3/Pack
Catalog No. 502830915
Encompass_Preferred
These 1/4-inch x 3-inch Steri-Strip™ adhesive skin closures contain polymer filaments for a secure closure They have a simple design that reduces scarring and chance of infection Made using a hypoallergenic adhesive, these 3M™ Steri-Strip™ reinforced skin closures are safe for sensitive skin A strong hold reduces tissue damage and allows for improved cosmetic results The breathable fabric stays gentle against the skin These non-woven reinforced strips are suitable for cuts, surgical incisions and after procedures requiring sutures and staples
Catalog No. 50-283-0915 Supplier Med Vet International Supplier No. 5780PK
These 1/4-inch x 3-inch Steri-Strip™ adhesive skin closures contain polymer filaments for a secure closure They have a simple design that reduces scarring and chance of infection Made using a hypoallergenic adhesive, these 3M™ Steri-Strip™ reinforced skin closures are safe for sensitive skin A strong hold reduces tissue damage and allows for improved cosmetic results The breathable fabric stays gentle against the skin These non-woven reinforced strips are suitable for cuts, surgical incisions and after procedures requiring sutures and staples