Learn More
Med Vet International Skin Closure Strip Steri-Strip 1/8 X 3 inch Nonwoven Material Reinforced Strip White, R1540, 5 Strips/Pack, 50 Packs/Box
Catalog No. 502830917
Encompass_Preferred
These 3M™ Steri-Strip™ reinforced skin closure strips close wounds, surgical incisions and lacerations They have reinforced filaments that provide extra strength These skin closures are made with a microporous material that helps to maintain skin health They reduce tissue trauma to minimize scarring These skin closures have pressure-sensitive adhesive to make application easy They're hypoallergenic and sterile
Catalog No. 50-283-0917 Supplier Med Vet International Supplier No. 5779BX
These 3M™ Steri-Strip™ reinforced skin closure strips close wounds, surgical incisions and lacerations They have reinforced filaments that provide extra strength These skin closures are made with a microporous material that helps to maintain skin health They reduce tissue trauma to minimize scarring These skin closures have pressure-sensitive adhesive to make application easy They're hypoallergenic and sterile