Learn More
Med Vet International Antimicrobial Skin Closure Strip Steri-Strip Antimicrobial 1/2 X 4 inch Nonwoven Material Reinforced Strip Tan, A1847, 6 Strips/Pack
Catalog No. 502830810
Encompass_Preferred
½ x 4 inches Reduces infection risk – Iodophor adhesive provides broad spectrum activity against the most common pathogens causing SSIs Stays in place – Excellent long-term hypoallergenic adhesion, to ensure wounds stay closed while healing Easy to apply – Fast application
Catalog No. 50-283-0810 Supplier Med Vet International Supplier No. 127107EA
½ x 4 inches Reduces infection risk – Iodophor adhesive provides broad spectrum activity against the most common pathogens causing SSIs Stays in place – Excellent long-term hypoallergenic adhesion, to ensure wounds stay closed while healing Easy to apply – Fast application