Learn More
Med Vet International Skin Closure Strip Steri-Strip 1/4 X 1-1/2 inch Nonwoven Material Reinforced Strip White, R1542, 6/Pack, 50 Packs/Box
Catalog No. 502830063
Encompass_Preferred
¼ Inch by 1½ Inches, reinforced Provides wound support and increases the tensile strength of the wound, compared to sutures Non-invasive, sterile design helps reduce scarring and the risk of infection, compared to sutures and staples, with less tissue trauma and better cosmetic outcomes Hypoallergenic adhesive Allows early staple/suture removal – Helps patients get back to their normal routine faster Easy to apply – Fast application
Catalog No. 50-283-0063 Supplier Med Vet International Supplier No. 5781BX
¼ Inch by 1½ Inches, reinforced Provides wound support and increases the tensile strength of the wound, compared to sutures Non-invasive, sterile design helps reduce scarring and the risk of infection, compared to sutures and staples, with less tissue trauma and better cosmetic outcomes Hypoallergenic adhesive Allows early staple/suture removal – Helps patients get back to their normal routine faster Easy to apply – Fast application