Learn More
Med Vet International 3M Steri-Strip Skin Closure Strip, 1/4 X 4in, 6 Strips/Envelope, 50 Envelopes/Box, R1546, 4 Boxes
Catalog No. 502832263
Encompass_Preferred
Skin Closure Strip Steri-Strip™ 1/4 X 4 Inch Nonwoven Material Reinforced Strip White¼ Inch by 4 Inches Provides wound support and increases the tensile strength of the wound, compared to -invasive, sterile design helps reduce scarring and the risk of infection, compared to sutures and staples, with less tissue trauma and better cosmetic outcomes Hypoallergenic adhesive Allows early staple/suture removal – Helps patients get back to their normal routine faster Easy to apply – Fast application
Catalog No. 50-283-2263 Supplier Med Vet International Supplier No. 5784CS
Skin Closure Strip Steri-Strip™ 1/4 X 4 Inch Nonwoven Material Reinforced Strip White¼ Inch by 4 Inches Provides wound support and increases the tensile strength of the wound, compared to -invasive, sterile design helps reduce scarring and the risk of infection, compared to sutures and staples, with less tissue trauma and better cosmetic outcomes Hypoallergenic adhesive Allows early staple/suture removal – Helps patients get back to their normal routine faster Easy to apply – Fast application