Learn More
Med Vet International Skin Closure Strip Steri-Strip 1/2 X 2 inch Nonwoven Material Reinforced Strip White, R1549, 6 Strips/Pack, 50 Packs/Box, 4 Boxes
Catalog No. 502830584
Encompass_Preferred
½ Inch by 2 Inches, reinforced Provides wound support and increases the tensile strength of the wound, compared to -invasive, sterile design helps reduce scarring and the risk of infection, compared to sutures and staples, with less tissue trauma and better cosmetic outcomes Hypoallergenic adhesive Allows early staple/suture removal – Helps patients get back to their normal routine faster Easy to apply – Fast application
Catalog No. 50-283-0584 Supplier Med Vet International Supplier No. 237687CS
½ Inch by 2 Inches, reinforced Provides wound support and increases the tensile strength of the wound, compared to -invasive, sterile design helps reduce scarring and the risk of infection, compared to sutures and staples, with less tissue trauma and better cosmetic outcomes Hypoallergenic adhesive Allows early staple/suture removal – Helps patients get back to their normal routine faster Easy to apply – Fast application