Learn More
Chemglass Life Sciences Pack, Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Amber with Grad Spot, 12 x 32mm, PTFE/Red Rubber Septa
SDP
Catalog No. 501532658
Encompass_Preferred
Pack, Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Amber with Grad Spot, 12 x 32mm, PTFE/Red Rubber Septa. These convenience packs come with 100 closures and vials packed seperately. Closures are not attached.
Catalog No. 50-153-2658 Supplier Chemglass Life Sciences Supplier No. CV53281233
Pack, Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Amber with Grad Spot, 12 x 32mm, PTFE/Red Rubber Septa. These convenience packs come with 100 closures and vials packed seperately. Closures are not attached.