Learn More
Chemglass Life Sciences Pack, Green Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear with Grad Spot, 12 x 32mm, PTFE/Silicone Septa
SDP
Catalog No. 501532651
Encompass_Preferred
Pack, Green Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear with Grad Spot, 12 x 32mm, PTFE/Silicone Septa. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Catalog No. 50-153-2651 Supplier Chemglass Life Sciences Supplier No. CV53231232G
Pack, Green Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear with Grad Spot, 12 x 32mm, PTFE/Silicone Septa. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.