Learn More
Thermo Scientific™ Closures with Prefitted Septa for 13mm Crimp/Snap-Ring LoVials™
Catalog No. 0339734 Shop All Thermo Scientific Products
Change view
Click to view available options
Septum:
Seal 2000, PTFE/RR
Catalog No. Septum
03-397-34 Seal 2000, PTFE/RR
1 options
Catalog No. 03-397-34 Supplier Thermo Scientific™ Supplier No. C401551
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
  • Kim-Snap™ closures provide a tight seal without the need of a crimper
  • National Scientific aluminum crimp closures provide a secure, leak-resistant seal
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
Septum Seal 2000, PTFE/RR
Closure Size 13 mm