Learn More
MilliporeSigma™ Supelco™ Closures for Snap Ring Vials
Catalog No. 111020946 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
11 mm Snap Ring
Wide Mouth Crimp/Snap Seal Vials
Material:
PTFE
PTFE, Silicone
No. per Pack:
100 Pk.
Septum:
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone, Starburst
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material No. per Pack Septum
11-102-0946 Wide Mouth Crimp/Snap Seal Vials PTFE, Silicone 100 Pk. PTFE/Silicone
11-101-1231 11 mm Snap Ring PTFE, Silicone 100 Pk. PTFE/Silicone, Starburst
11-101-1373 11 mm Snap Ring PTFE, Silicone 100 Pk. PTFE/Silicone, Pre-slit
11-101-2016 Wide Mouth Crimp/Snap Seal Vials PTFE 100 Pk. -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-0946 Supplier MilliporeSigma™ Supelco™ Supplier No. 24758
Add to Cart
Add to Cart
Closure type snap ring seal
  • Closure type snap ring seal
  • For snap ring vials
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Opening 11 mm
Material PTFE, Silicone
Pre-slit No
Septum PTFE/Silicone
Material (Seal) Polypropylene
No. per Pack 100 Pk.
For Use With (Equipment) Wide Mouth Crimp/Snap Seal Vials