Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Closures for Snap Ring Vials with Snap Cap

Catalog No. 111012033 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Polyethylene, PTFE, Rubber
Polyethylene, PTFE, Silicone
Hardness:
45 (Durometer: shore A)
60 (Durometer: shore A)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Hardness
11-101-2033 Polyethylene, PTFE, Rubber 60 (Durometer: shore A)
11-101-1911 Polyethylene, PTFE, Silicone 45 (Durometer: shore A)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2033 Supplier MilliporeSigma™ Supelco™ Supplier No. SU860090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Closure type clear polyethylene snap cap

  • Closure type clear polyethylene snap cap
  • Compatible with Large Opening Crimp/Snap Seal Vials
  • Clear polyethylene cap
TRUSTED_SUSTAINABILITY

Specifications

Description Snap cap
Color Red, Clear
Diameter (Metric) Outer 11.5 mm
Material Polyethylene, PTFE, Rubber
Septum PTFE/Red Rubber
For Use With (Application) 11 mm Snap Ring
No. per Pack 100
Hardness 60 (Durometer: shore A)
Quantity 100 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.