Learn More
Med Vet International Clot Blood Collection Tube, Red Cap, 3ml, J1561, Each
Catalog No. 5000583077
Encompass_Preferred
JorVet offers a wide range of blood collection tubes in various sizes and formats. All tubes adhere to the high standards of human blood collection tubes. Plastic versus Glass Human blood collection tubes have generally switched over to PET plastic tubes versus breakable glass. JorVet also uses the latest standardized color plastic caps for consistency. All tubes are in storage tray of 100 tubes Red Tubes are clot tubes where serum analysis is desired. Clot Blood Collection Tube - 3ml
Catalog No. 50-005-83077 Supplier Med Vet International Supplier No. JOR1561
JorVet offers a wide range of blood collection tubes in various sizes and formats. All tubes adhere to the high standards of human blood collection tubes. Plastic versus Glass Human blood collection tubes have generally switched over to PET plastic tubes versus breakable glass. JorVet also uses the latest standardized color plastic caps for consistency. All tubes are in storage tray of 100 tubes Red Tubes are clot tubes where serum analysis is desired. Clot Blood Collection Tube - 3ml