Learn More
Nova Biomedical Corporation CLOT CATCHER 38883 200/PK
Catalog No. nc0249531
Encompass
Capillary Clot Catchers, by Nova Biomedical, are designed to prevent clots present in whole blood samples from entering the flowpath of Nova Stat Profile pHOx series and Critical Care Xpress analyzers. These disposable, single-use devices are particularly beneficial when processing difficult to obtain neonatal and pediatric samples.
Catalog No. NC0249531 Supplier Nova Biomedical Corporation Supplier No. 38883
Capillary Clot Catchers, by Nova Biomedical, are designed to prevent clots present in whole blood samples from entering the flowpath of Nova Stat Profile pHOx series and Critical Care Xpress analyzers. These disposable, single-use devices are particularly beneficial when processing difficult to obtain neonatal and pediatric samples.