Learn More
Uline Cloth Parts Bags - 5 x 7, White
Catalog No. NC1521836
Encompass_Preferred
100% cotton, Weave Count 20 x 20, Single Drawstring, Not treated with chemicals, Not pre-shrunk Will shrink up to 10%, Small red stitch at the top of the bag, Not FDA compliant
Catalog No. NC1521836 Supplier Uline Supplier No. S1164PK
100% cotton, Weave Count 20 x 20, Single Drawstring, Not treated with chemicals, Not pre-shrunk Will shrink up to 10%, Small red stitch at the top of the bag, Not FDA compliant