Learn More
Chem Service Inc Clothianidin Solution,210880-92-5,1ML
SDP
Catalog No. NC9858025
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
100ug/ml in Acetonitrile; Boiling Point 81.6 C; Melting Point -45 C; Flash Point 5.6 C
Catalog No. NC9858025 Supplier Chem Service Inc Supplier No. S11493A11ML
This item has a minimum order quantity of 2 per supplier requirements
100ug/ml in Acetonitrile; Boiling Point 81.6 C; Melting Point -45 C; Flash Point 5.6 C