Learn More
Medchemexpress LLC Clothianidin-d3 | 1262776-24-8 | MFCD17019117 | 252.70 g/mol | C6H8ClN5O2S | 5 MG
SDP
Catalog No. 5000262408
Encompass_Preferred
Clothianidin-d3 is a deuterium-labeled analogue of the neonicotinoid insecticide clothianidin, provided as a 5 mg analytical reference standard for use as an internal standard or reference in mass spectrometry, residue, and trace analysis.
  • Isotopically labeled with three deuterium atoms (d3) for accurate mass spectrometry quantification.
  • Intended for use as an analytical reference standard and internal standard in LC-MS/MS or GC-MS analyses.
  • Provided as a small, neat solid suitable for preparing calibration standards and spike recoveries.
  • CAS 1262776-24-8 identifies the deuterated compound for reference in analytical workflows.
  • Useful for pesticide residue analysis and trace-level quantification in environmental and food matrices.
Catalog No. 50-002-62408 Supplier Medchemexpress LLC Supplier No. HY133167S5MG
Clothianidin-d3 is a deuterium-labeled analogue of the neonicotinoid insecticide clothianidin, provided as a 5 mg analytical reference standard for use as an internal standard or reference in mass spectrometry, residue, and trace analysis.
  • Isotopically labeled with three deuterium atoms (d3) for accurate mass spectrometry quantification.
  • Intended for use as an analytical reference standard and internal standard in LC-MS/MS or GC-MS analyses.
  • Provided as a small, neat solid suitable for preparing calibration standards and spike recoveries.
  • CAS 1262776-24-8 identifies the deuterated compound for reference in analytical workflows.
  • Useful for pesticide residue analysis and trace-level quantification in environmental and food matrices.