Learn More
AdipoGen Clothianidin-d3
SDP
Supplier:  AdipoGen CDXC0190M100
Encompass_Preferred
Chemical. CAS 1262776-24-8. Formula C6D3H5ClN5O2S. MW 252.7. Clothianidin-d3 is deuterium labelled Clothianidin, which is a neonicotinoid insecticide. Neonicotinoids NNIs are a family of active substances used in plant protection to control harmful insects and also in veterinary medicine. Neonicotinoids act selectively on the insect nicotinic acetylcholine receptors nAChRs, initially stimulating postsynaptic receptors, increasing Na ingress and K egress and then paralyzing nerve conduction, resulting in rapid death. This compound can be used as a reference compound, in the determination of several neonicotinoid insecticides present in bee pollen, using ultra-high performance liquid chromatography UHPLC coupled with mass spectrometry MS.
Catalog No. 50-256-1429