Learn More
Enzo Life Sciences Clothiapine (20mg). CAS: 2058-52-8
SDP
Supplier:  Enzo Life Sciences BMLD1080020
Encompass_Preferred
Atypical neuroleptic agent related to clozapine. Purity: ≥99%. Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0494