Learn More
Cayman Chemical Clotrimazole, Purity: ≥98%, CAS: 23593-75-1, 100 mg
SDP
Catalog No. 502971834
Encompass_Preferred
An imidazole antifungal; tightly binds sterol 14-α demethylase isoform B from A. fumigatus (KD = 103 nM) and disturbs the fungal cell membrane; inhibits the calcium-dependent potassium channels Kv1.3 and IK-1 (IC50 = 6.0 and 0.07 µM, respectively) in mammalian cells
Catalog No. 50-297-1834 Supplier Cayman Chemical Supplier No. 15278100 MG
An imidazole antifungal; tightly binds sterol 14-α demethylase isoform B from A. fumigatus (KD = 103 nM) and disturbs the fungal cell membrane; inhibits the calcium-dependent potassium channels Kv1.3 and IK-1 (IC50 = 6.0 and 0.07 µM, respectively) in mammalian cells