Learn More
Medchemexpress LLC Clotrimazole | 23593-75-1 | 99.91% | 344.84 | 5 G
SDP
Catalog No. 5000346017
Encompass_Preferred
Clotrimazole is an imidazole derivative and an antifungal compound. It acts as a CYP (cytochrome P450) inhibitor and also exhibits antibacterial activity, making it suitable for studying fungal infections such as vaginal yeast infections, oral thrush, and ringworm.
  • Antifungal properties
  • CYP (cytochrome P450) inhibitor
  • Antibacterial activity
  • Useful for research into fungal infections
Catalog No. 50-003-46017 Supplier Medchemexpress LLC Supplier No. HY108821G
Clotrimazole is an imidazole derivative and an antifungal compound. It acts as a CYP (cytochrome P450) inhibitor and also exhibits antibacterial activity, making it suitable for studying fungal infections such as vaginal yeast infections, oral thrush, and ringworm.
  • Antifungal properties
  • CYP (cytochrome P450) inhibitor
  • Antibacterial activity
  • Useful for research into fungal infections