Learn More
Cayman Chemical ClotrImazole 50mg
SDP
Catalog No. 502971321
Encompass_Preferred
An imidazole antifungal; tightly binds sterol 14-α demethylase isoform B from A. fumigatus (KD = 103 nM) and disturbs the fungal cell membrane; inhibits the calcium-dependent potassium channels Kv1.3 and IK-1 (IC50 = 6.0 and 0.07 µM, respectively) in mammalian cells
Catalog No. 50-297-1321 Supplier Cayman Chemical Supplier No. 1527850 MG
An imidazole antifungal; tightly binds sterol 14-α demethylase isoform B from A. fumigatus (KD = 103 nM) and disturbs the fungal cell membrane; inhibits the calcium-dependent potassium channels Kv1.3 and IK-1 (IC50 = 6.0 and 0.07 µM, respectively) in mammalian cells