Learn More
Med Vet International Clotrimazole With Betamethasone Cream, 15gm, Generic Lotrisone
Catalog No. 502828107
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Clotrimazole with Betamethasone cream is a generic version of Lotrisone. This product contains 2 medication, Clotrimazole which is an azole antifungal and Betamethasone which is a strong corticosteroid. This product should be stored at room temperature away from light and moisture.
Catalog No. 50-282-8107 Supplier Med Vet International Supplier No. RX51672404801
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Clotrimazole with Betamethasone cream is a generic version of Lotrisone. This product contains 2 medication, Clotrimazole which is an azole antifungal and Betamethasone which is a strong corticosteroid. This product should be stored at room temperature away from light and moisture.