Learn More
Medchemexpress LLC Clotrimazole-d10 | 23593-75-1 | 97.5% | 354.90 | 25MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10882S1
Encompass_Preferred
Clotrimazole-d10 is deuterated labeled Clotrimazole. Clotrimazole is an imidazole derivative, an antifungal compound, and a CYP (cytochrome P450) inhibitor. It also possesses antibacterial activity.
  • Deuterated labeled clotrimazole
  • Imidazole derivative
  • Antifungal compound
  • CYP (cytochrome P450) inhibitor
  • Antibacterial activity
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-47740