Learn More
Medchemexpress LLC Clotrimazole-d5 | 1185076-41-8 | 99.47% | 349.87 | 1 MG
SDP
Catalog No. 5000345833
Encompass_Preferred
This product is a deuterium-labeled form of an imidazole derivative. It functions as an antifungal compound and an inhibitor of cytochrome P450 enzymes. This compound also exhibits antibacterial activity and is primarily used for research purposes.
  • Functions as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Exhibits antifungal properties.
  • Acts as an inhibitor of cytochrome P450.
  • Possesses antibacterial activity.
Catalog No. 50-003-45833 Supplier Medchemexpress LLC Supplier No. HY10882S1MG
This product is a deuterium-labeled form of an imidazole derivative. It functions as an antifungal compound and an inhibitor of cytochrome P450 enzymes. This compound also exhibits antibacterial activity and is primarily used for research purposes.
  • Functions as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Exhibits antifungal properties.
  • Acts as an inhibitor of cytochrome P450.
  • Possesses antibacterial activity.