Learn More
Abcam Clotrimazole, Non-selective P450 cytochrome inhibitor, 50MG
Catalog No. 502707311
Encompass_Preferred
MW 344.84 g/mol, Purity >99%. Non-selective P450 cytochrome inhibitor (Ki = 0.02-67 μM for P450 cyctochrome subtypes). Potent inhibitor of 34A (Ki = 20 nM). Antifungal agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7311 Supplier Abcam Supplier No. AB12074450MG
MW 344.84 g/mol, Purity >99%. Non-selective P450 cytochrome inhibitor (Ki = 0.02-67 μM for P450 cyctochrome subtypes). Potent inhibitor of 34A (Ki = 20 nM). Antifungal agent.
The product is subject to the following: Abcam Restricted Use Statement