Learn More
AccuStandard, Inc Cloud Point Standard | Approx. -10C | 250ml
SDP
Catalog No. NC1564112
Encompass
This measures the temperature at which wax (paraffin) begins to separate when oil chilled to a low temperature, and it serves as an important indicator of practical performance in automotive applications in low temperatures.
Catalog No. NC1564112 Supplier AccuStandard, Inc Supplier No. ASTMP13103
This measures the temperature at which wax (paraffin) begins to separate when oil chilled to a low temperature, and it serves as an important indicator of practical performance in automotive applications in low temperatures.