Learn More
Pharmacal Research Laboratories CLOUT 30GALLON
SDP
Catalog No. NC0071004
Encompass
Clout� is a high detergency, low alkaline pH cage and rack washing compound. Clout� has been specifically formulated to clean without the use of caustic alkaline ingredients. This product is safe for use with polycarbonate caging materials, rubber, glass and stainless steel. 30 Gallon drum.
Catalog No. NC0071004 Supplier Pharmacal Research Laboratories Supplier No. 18030F
Clout� is a high detergency, low alkaline pH cage and rack washing compound. Clout� has been specifically formulated to clean without the use of caustic alkaline ingredients. This product is safe for use with polycarbonate caging materials, rubber, glass and stainless steel. 30 Gallon drum.