Learn More
Pharmacal Research Laboratories Clout - High Detergency, Low Alkaline pH, Safe for Various Materials, 55 Gallon Drum
SDP
Catalog No. NC0536326
Encompass
CLOUT - 55 Gallon Drum is a high-detergency, low alkaline pH washing compound, formulated to clean efficiently without harsh alkaline ingredients. It's compatible with diverse materials like polycarbonate, rubber, glass, and stainless steel, making it ideal for mechanical cage washers. With a pH of 12.5 and a 3-year storage stability, this product offers effective cleaning while ensuring safety for various materials.
Catalog No. NC0536326 Supplier Pharmacal Research Laboratories Supplier No. 18055F
CLOUT - 55 Gallon Drum is a high-detergency, low alkaline pH washing compound, formulated to clean efficiently without harsh alkaline ingredients. It's compatible with diverse materials like polycarbonate, rubber, glass, and stainless steel, making it ideal for mechanical cage washers. With a pH of 12.5 and a 3-year storage stability, this product offers effective cleaning while ensuring safety for various materials.