Learn More
Pharmacal Research Laboratories CLOUT ALKALINE DETERGENT 5GAL
SDP
Catalog No. NC9643002
Encompass
Clout Alkaline Detergent, Is a high detergency, low alkaline pH cage and rack washing compound, Has been specifically formulated to clean without the use of caustic alkaline ingredients, Size: 5 gal. Cube
Catalog No. NC9643002 Supplier Pharmacal Research Laboratories Supplier No. 18025F
Clout Alkaline Detergent, Is a high detergency, low alkaline pH cage and rack washing compound, Has been specifically formulated to clean without the use of caustic alkaline ingredients, Size: 5 gal. Cube