Learn More
Pharmacal Research Laboratories Clout Aqua 5 Gal, Alkaline Aquatic Tank Cleaner, Removes Algae And Biofilm, pH 13, 5 GAL
SDP
Supplier:  Pharmacal Research Laboratories 19525F
Encompass
CLOUT AQUA 5 GAL is a powerful alkaline cleaning compound specifically designed for aquatic tanks, effectively removing algae and biofilm. With a pH of 13, it offers professional-grade cleaning without the risk of flammability. This clear, colorless liquid is intended for professional use and is safe to store in well-ventilated areas, ensuring high efficacy and safety for aquatic environments.
Catalog No. NC2104933