Learn More
Medchemexpress LLC Clovibactin TFA | 97.1% | 903.08 | 1 MG
SDP
Catalog No. 5000425064
Encompass_Preferred
Clovibactin TFA is the TFA salt form of Clovibactin (HY-P10027), serving as an antibiotic for drug-resistant bacterial pathogens without detectable resistance. It functions by inhibiting cell wall synthesis, specifically targeting pyrophosphate of peptidoglycan precursors.
  • Effective against drug-resistant bacterial pathogens.
  • No detectable resistance observed.
  • Inhibits cell wall synthesis through a specific mechanism targeting peptidoglycan precursors.
  • Demonstrates inhibition of Staphylococcus strains with MICs ranging from 0.125-2 μg/mL in vitro.
  • Exhibits a characterized pharmacokinetic profile in mice, including a Cmax of 219.3 μg/mL and a t1/2 of 2 hours following a 20 mg/kg intravenous single dose.
Catalog No. 50-004-25064 Supplier Medchemexpress LLC Supplier No. HYP10027A1MG
Clovibactin TFA is the TFA salt form of Clovibactin (HY-P10027), serving as an antibiotic for drug-resistant bacterial pathogens without detectable resistance. It functions by inhibiting cell wall synthesis, specifically targeting pyrophosphate of peptidoglycan precursors.
  • Effective against drug-resistant bacterial pathogens.
  • No detectable resistance observed.
  • Inhibits cell wall synthesis through a specific mechanism targeting peptidoglycan precursors.
  • Demonstrates inhibition of Staphylococcus strains with MICs ranging from 0.125-2 μg/mL in vitro.
  • Exhibits a characterized pharmacokinetic profile in mice, including a Cmax of 219.3 μg/mL and a t1/2 of 2 hours following a 20 mg/kg intravenous single dose.