Learn More
Apexbio Technology LLC Cloxacillin Sodium 7081-44-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092721
Encompass_Preferred
Cloxacillin Sodium (CAS 7081-44-9) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall biosynthesis thereby disrupting peptidoglycan cross-linking and inducing bacterial cell death Cloxacillin Sodium exerts its biological activity primarily through inhibition of PBPs leading to inactivation of cell wall synthesis in bacteria In in vitro studies Cloxacillin Sodium demonstrates inhibitory activity with an IC50 value of 0 04 M depending on the bacterial strain and experimental conditions Based on these pharmacological properties Cloxacillin Sodium holds research potential in studying bacterial pathogen susceptibility antibiotic resistance mechanisms and evaluation of antibiotic efficacy
Catalog No. 50-000-92721 Supplier Apexbio Technology LLC Supplier No. B19185.1
Cloxacillin Sodium (CAS 7081-44-9) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall biosynthesis thereby disrupting peptidoglycan cross-linking and inducing bacterial cell death Cloxacillin Sodium exerts its biological activity primarily through inhibition of PBPs leading to inactivation of cell wall synthesis in bacteria In in vitro studies Cloxacillin Sodium demonstrates inhibitory activity with an IC50 value of 0 04 M depending on the bacterial strain and experimental conditions Based on these pharmacological properties Cloxacillin Sodium holds research potential in studying bacterial pathogen susceptibility antibiotic resistance mechanisms and evaluation of antibiotic efficacy