Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cloxacillin sodium monohydrate

Catalog No. AAJ6650003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6650003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6650003 Supplier Thermo Scientific Chemicals Supplier No. J6650003

missing translation for 'validMainframeMsg'

CAS: 7081-44-9 | C19H19ClN3NaO6S | 475.876 g/mol

Targets primarily Gram-positive bacteria, especially --lactamase producing Staphylococci

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Targets primarily gram positive bacteria, especially _x001A_-lactamase producing Staphylococci
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7081-44-9
Molecular Formula C19H19ClN3NaO6S
Molecular Weight (g/mol) 475.876
MDL Number MFCD00150735
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
PubChem CID 23675743
ChEBI CHEBI:34978
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]

Specifications

Type Cloxacillin Sodium Monohydrate
Physical Form Powder
Melting Point 170°C
Color White
Quantity 1 g
Flash Point >110°C (230°F)
Boiling Point 689°C
Merck Index 14,2419
Solubility Information Soluble in water
Formula Weight 475.88
Percent Purity 95%
Chemical Name or Material Cloxacillin sodium monohydrate
Show More Show Less
Hazard Category H315-H317-H319-H334-H335
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 211-390-9
RTECSNumber XH8920000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.