Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cloxacillin sodium salt monohydrate

Catalog No. AC455300010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455300010 1 g
AC455300050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC455300010 Supplier Thermo Scientific Chemicals Supplier No. 455300010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Cloxacillin sodium salt monohydrate, CAS # 7081-44-9, is a compound that targets gram-positive bacteria and shows antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cloxacillin sodium salt monohydrate is a chlorinated derivative of oxacillin that shows β-lactamase-resistant antibiotic activity

• This compound inhibits the last stage of bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), which results in cell lysis, that is mediated by bacterial cell wall autolytic enzymes

Applications

• Cloxacillin sodium salt monohydrate targets gram-positive bacteria, and is effective against staphylococci that produce β-lactamase

• It acts in vitro against both penicillin gram-resistant and gram-sensitive staphylococci, certain streptococci, and pneumococci

• This compound is commonly used in vivo and in vitro studies as AmpC β-lactamase inhibitor in combination with β-lactam antibiotics

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7081-44-9
Molecular Formula C19H19ClN3NaO6S
Molecular Weight (g/mol) 475.876
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
PubChem CID 23675743
ChEBI CHEBI:34978
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]

Specifications

Type Cloxacillin Sodium Salt Monohydrate
Infrared Spectrum Authentic
Water 3.0 to 5.0%
Quantity 1 g
Formula Weight 475.88
Percent Purity ≥94% (HPLC)
Chemical Name or Material Cloxacillin sodium salt monohydrate
Hazard Category Danger
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.