Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cloxacillin Sodium Salt, MP Biomedicals™

Catalog No. MP215070225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215070225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215070225 Supplier MP Biomedicals, Inc Supplier No. 0215070225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs). Cloxacillin Sodium Salt, MP Biomedicals is a semi-synthetic, β-lactamase-resistant antibiotic used to treat infections. It is a chlorin

Cloxacillin binds to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. It may interfere with autolysin inhibitors. Cloxacillin has many antibiotic uses:

  • To treat infections caused by penicillin G-resistant staphylococci
  • In studies of the use of topical antibacterial agents for burn and crush wounds
  • As an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7081-44-9
Molecular Formula C19H19ClN3NaO6S
Molecular Weight (g/mol) 475.876
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
Synonym 5-Methyl-3-(o-chlorophenyl)-4-isoxazolyl] pencillin, cloxacillin sodium
PubChem CID 23675743
ChEBI CHEBI:34978
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]

Specifications

Physical Form Powder
pH 6.0 to 7.5 (literature)
Color White to Off-White
Quantity 25 g
Solubility Information Freely soluble in water (200mg + 4 mL, clear colorless to faint yellow solution) and methanol; soluble in alcohol, ethylene gycol, ethyl, and pyridene.
Formula Weight 457.86
Percent Purity 100.8%
Grade Not Available
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.