Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cloxacillin sodium salt

Catalog No. AAJ6431414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6431414 25 g
AAJ6431403 1 g
AAJ6431406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6431414 Supplier Thermo Scientific Chemicals Supplier No. J6431414
Add to Cart

A semisynthetic antibiotic | CAS: 642-78-4 | C19H18ClN3NaO5S | 458.869 g/mol

Cloxacillin sodium salt is used to treat infections caused by penicillin G-resistant staphylococci. It has been used in studies of topical antibacterial agent for burn and crush wounds. It is used as an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cloxacillin sodium salt is used to treat infections caused by penicillin G-resistant staphylococci. It has been used in studies of topical antibacterial agent for burn and crush wounds. It is used as an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-78-4
Molecular Formula C19H18ClN3NaO5S
Molecular Weight (g/mol) 458.869
MDL Number MFCD00063568
InChI Key JBJAWVZHLGPBJA-SLINCCQESA-N
PubChem CID 66587017
IUPAC Name (2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na]

Specifications

Type Cloxacillin Sodium Salt
Physical Form Powder
Melting Point 170°C
Color White
Quantity 25 g
Beilstein 4103180
Merck Index 14,2419
Solubility Information Soluble in water
Formula Weight 457.86
Chemical Name or Material Cloxacillin sodium salt
EINECSNumber 211-390-9
RTECSNumber XH8750000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.