Learn More
Medchemexpress LLC Cloxiquine | 130-16-5 | 99.9% | 10 G
SDP
Catalog No. 5000407857
Encompass_Preferred
Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal, and antiamoebic agent used in the research of tuberculosis and dermatoses. It has been shown to suppress the growth and metastasis of melanoma cells by activating PPARγ. This compound appears as a light yellow to yellow solid.
  • Antibacterial, antifungal, and antiamoebic agent
  • Used in tuberculosis and dermatoses research
  • Suppresses melanoma cell growth and metastasis through PPARγ activation
  • Exhibits antituberculosis activities
  • Inhibits B16F10 and A375 cell growth and migration
  • Suppresses glycolysis in B16F10 cells
  • Suppresses tumor growth and metastasis in mouse melanoma models
Catalog No. 50-004-07857 Supplier Medchemexpress LLC Supplier No. HYB096310G
Add to Cart
Add to Cart
Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal, and antiamoebic agent used in the research of tuberculosis and dermatoses. It has been shown to suppress the growth and metastasis of melanoma cells by activating PPARγ. This compound appears as a light yellow to yellow solid.
  • Antibacterial, antifungal, and antiamoebic agent
  • Used in tuberculosis and dermatoses research
  • Suppresses melanoma cell growth and metastasis through PPARγ activation
  • Exhibits antituberculosis activities
  • Inhibits B16F10 and A375 cell growth and migration
  • Suppresses glycolysis in B16F10 cells
  • Suppresses tumor growth and metastasis in mouse melanoma models