Learn More
Medchemexpress LLC Cloxiquine | 130-16-5 | 99.9% | 100 G
SDP
Catalog No. 5000408343
Encompass_Preferred
Cloxiquine (5-Chloro-8-quinolinol) is a versatile agent with antibacterial, antifungal, and antiamoebic properties. It is suitable for research concerning tuberculosis and dermatoses. This compound has demonstrated the ability to suppress the growth and metastasis of melanoma cells by activating PPARγ, and also exhibits antituberculosis activities against various strains of *Mycobacterium tuberculosis*.
  • Antibacterial, antifungal, and antiamoebic agent.
  • Used for tuberculosis and dermatoses research.
  • Suppresses melanoma cell growth and metastasis by activating PPARγ.
  • Exhibits antituberculosis activities with MICs ranging from 0.062 to 0.25 μg/mL.
  • Inhibits migration of B16F10 and A375 cells.
  • Suppresses glycolysis in B16F10 cells.
  • Suppresses tumor growth in mouse melanoma xenograft model.
  • Suppresses tumor metastasis in mouse melanoma lung metastatic model.
Catalog No. 50-004-08343 Supplier Medchemexpress LLC Supplier No. HYB0963100G
Cloxiquine (5-Chloro-8-quinolinol) is a versatile agent with antibacterial, antifungal, and antiamoebic properties. It is suitable for research concerning tuberculosis and dermatoses. This compound has demonstrated the ability to suppress the growth and metastasis of melanoma cells by activating PPARγ, and also exhibits antituberculosis activities against various strains of *Mycobacterium tuberculosis*.
  • Antibacterial, antifungal, and antiamoebic agent.
  • Used for tuberculosis and dermatoses research.
  • Suppresses melanoma cell growth and metastasis by activating PPARγ.
  • Exhibits antituberculosis activities with MICs ranging from 0.062 to 0.25 μg/mL.
  • Inhibits migration of B16F10 and A375 cells.
  • Suppresses glycolysis in B16F10 cells.
  • Suppresses tumor growth in mouse melanoma xenograft model.
  • Suppresses tumor metastasis in mouse melanoma lung metastatic model.