Learn More
Cayman Chemical ClozapIn-d4 500ug
SDP
Catalog No. 502980023
Encompass_Preferred
An internal standard for the quantification of clozapine by GC- or LC-MS
Catalog No. 50-298-0023 Supplier Cayman Chemical Supplier No. 17513500 UG
An internal standard for the quantification of clozapine by GC- or LC-MS